Anti-ATP1A3 Mouse Monoclonal Antibody [clone: XVIF9-G10]
Catalog # ABCA_AB2826-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:ATP1A3_DOG
- Antigen Symbol:ATP1A3
- Clonality:Monoclonal
- Clone:XVIF9-G10
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:RDP|Alpha(III)|Sodium pump 3|Na+/K+ ATPase 3|Sodium pump subunit alpha-3|Na+/K+ ATPase alpha 3 subunit|Na(+)/K(+) ATPase alpha-3 subunit|Na(+)/K(+) ATPase alpha(III) subunit|Sodium potassium ATPase alpha 3 polypeptide|ATPase Na+/K+ transporting alpha 3 polypeptide|Sodium/potassium transporting ATPase alpha 3 chain|Sodium/potassium-transporting ATPase subunit alpha-3
- Storage Buffer:PBS
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [XVIF9-G10] to ATP1A3.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATP1A3
Clonality: Monoclonal
Clone: XVIF9-G10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse; Rat; Human