Anti-MIP-1 alpha/CCL3 Rabbit Polyclonal Antibody
Supplier: Abcam
New Product
Certificates
About this item
Rabbit polyclonal to MIP-1 alpha/CCL3.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MIP-1 alpha/CCL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB32609-1ML
- ABCA_AB32609-500UL
- Antigen Symbol:
- MIP-1 alpha/CCL3
- MIP-1 alpha/CCL3
- Antigen Name:
- G0S19-1
- G0S19-1
- Antigen Synonyms:
- MIP1A|SCYA3|CCL3|C-C motif chemokine 3|G0/G1 switch regulatory protein 19-1|Macrophage inflammatory protein 1-alpha|PAT 464.1|SIS-beta|Small-inducible cytokine A3|Tonsillar lymphocyte LD78 alpha protein|MIP-1-alpha
- MIP1A|SCYA3|CCL3|C-C motif chemokine 3|G0/G1 switch regulatory protein 19-1|Macrophage inflammatory protein 1-alpha|PAT 464.1|SIS-beta|Small-inducible cytokine A3|Tonsillar lymphocyte LD78 alpha protein|MIP-1-alpha
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 10085 Da
- 10085 Da
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Storage Buffer:
- PBS, BSA
- PBS, BSA
- Storage Temperature:
- Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P
Specifications
Frequently Bought Together