Order Entry
United States
ContactUsLinkComponent
Anti-Drosophila FMR1 Mouse Monoclonal Antibody [clone: 6A15]
Anti-Drosophila FMR1 Mouse Monoclonal Antibody [clone: 6A15]
Catalog # ABCA_AB10299-100UL
Supplier:  Abcam
New Product
Anti-Drosophila FMR1 Mouse Monoclonal Antibody [clone: 6A15]
Catalog # ABCA_AB10299-100UL
Supplier:  Abcam
Supplier Number:  AB10299-100UL
New Product

About this item

Mouse monoclonal [6A15] to Drosophila FMR1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Drosophila FMR1
Clonality: Monoclonal
Clone: 6A15
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Drosophila melanogaster