Anti-KIR2DL3 Rabbit Monoclonal Antibody [clone: EPR22195]
: Abcam
New Product
Rabbit monoclonal [EPR22195] to KIR2DL3.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: KIR2DL3
Clonality: Monoclonal
Clone: EPR22195
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB224698-100U
- ABCA_AB224698-1ML
- Antigen Symbol:
- KIR2DL3
- KIR2DL3
- Antigen Name:
- CD158b2
- CD158b2
- Antigen Synonyms:
- KIRCL23|NKAT2|Killer cell immunoglobulin-like receptor 2DL3|CD158 antigen-like family member B2|KIR-023GB|Killer inhibitory receptor cl 2-3|NKAT2a|NKAT2b|Natural killer-associated transcript 2|p58 natural killer cell receptor clone CL-6|p58.2 MHC class-I-specific NK receptor|NKAT-2|p58 NK receptor CL-6
- KIRCL23|NKAT2|Killer cell immunoglobulin-like receptor 2DL3|CD158 antigen-like family member B2|KIR-023GB|Killer inhibitory receptor cl 2-3|NKAT2a|NKAT2b|Natural killer-associated transcript 2|p58 natural killer cell receptor clone CL-6|p58.2 MHC class-I-specific NK receptor|NKAT-2|p58 NK receptor CL-6
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR22195
- EPR22195
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 37886 Da
- 37886 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
Frequently Bought Together