Anti-HIF1 beta Mouse Monoclonal Antibody [clone: 29/HIF-1b]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [29/HIF-1b] to HIF1 beta - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HIF1 beta
Clonality: Monoclonal
Clone: 29/HIF-1b
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse
Specifications
- Catalog No:
- ABCA_AB289560-100U
- ABCA_AB289560-1MG
- Antigen Symbol:
- HIF1 beta
- HIF1 beta
- Antigen Name:
- BHLHE2
- BHLHE2
- Antigen Synonyms:
- ARNT|Aryl hydrocarbon receptor nuclear translocator|ARNT protein|Class E basic helix-loop-helix protein 2|Hypoxia-inducible factor 1-beta|HIF-1-beta|HIF1-beta
- ARNT|Aryl hydrocarbon receptor nuclear translocator|ARNT protein|Class E basic helix-loop-helix protein 2|Hypoxia-inducible factor 1-beta|HIF-1-beta|HIF1-beta
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 29/HIF-1b
- 29/HIF-1b
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 86636 Da
- 86636 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- ICC/IF
- ICC/IF
Specifications
Frequently Bought Together