Anti-YKL-40/CHI3L1 Rabbit Recombinant Multiclonal Antibody [clone: RM1204]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit recombinant multiclonal [RM1204] to YKL-40/CHI3L1 - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: YKL-40/CHI3L1
Clonality: Recombinant Multiclonal
Clone: RM1204
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB319165-1MG
- ABCA_AB319165-100U
- Antigen Symbol:
- YKL-40/CHI3L1
- YKL-40/CHI3L1
- Antigen Name:
- Chitinase-3-like protein 1
- Chitinase-3-like protein 1
- Antigen Synonyms:
- 39 kDa synovial protein|Cartilage glycoprotein 39|YKL-40|CGP-39|GP-39|hCGP-39|CHI3L1
- 39 kDa synovial protein|Cartilage glycoprotein 39|YKL-40|CGP-39|GP-39|hCGP-39|CHI3L1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Multiclonal
- Recombinant Multiclonal
- Clone:
- RM1204
- RM1204
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 42625 Da
- 42625 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra), IHC-P
- Flow Cyt (Intra), IHC-P
Specifications
Frequently Bought Together