Anti-TRRAP Rabbit Monoclonal Antibody [clone: EPR14952]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR14952] to TRRAP.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TRRAP
Clonality: Monoclonal
Clone: EPR14952
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB183517-1ML
- ABCA_AB183517-100U
- Antigen Symbol:
- TRRAP
- TRRAP
- Antigen Name:
- PAF400
- PAF400
- Antigen Synonyms:
- Transformation/transcription domain-associated protein|350/400 kDa PCAF-associated factor|STAF40|Tra1 homolog|PAF350/400
- Transformation/transcription domain-associated protein|350/400 kDa PCAF-associated factor|STAF40|Tra1 homolog|PAF350/400
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR14952
- EPR14952
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 437600 Da
- 437600 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Concentration:
- 0.536 mg/ml
- 0.536 mg/ml
- Storage Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- ICC/IF
- ICC/IF
Specifications
Frequently Bought Together