Anti-STAT5b (phospho S731) Rabbit Polyclonal Antibody
Catalog # ABCA_AB52211-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Signal transducer and activator of transcription 5B
- Antigen Symbol:STAT5b
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Modification:phospho S731
- Storage Buffer:PBS, Glycerol, Sodium chloride
- Molecular Weight:89866 Da
- Storage Temperature:Store at 4 °C (up to 6 months). Store at –20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human STAT5B phospho S731 aa 650 - 750. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to STAT5b (phospho S731).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: STAT5b
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human