Anti-SIRP alpha Mouse Monoclonal Antibody [clone: SADF10.1]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [SADF10.1] to SIRP alpha.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SIRP alpha
Clonality: Monoclonal
Clone: SADF10.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB267409-100U
- ABCA_AB267409-1MG
- Antigen Symbol:
- SIRP alpha
- SIRP alpha
- Antigen Name:
- CD172a
- CD172a
- Antigen Synonyms:
- BIT|MFR|MYD1|PTPNS1|SHPS1|SIRP|SIRPA|Tyrosine-protein phosphatase non-receptor type substrate 1|SHP substrate 1|SHPS-1|Brain Ig-like molecule with tyrosine-based activation motifs|CD172 antigen-like family member A|Inhibitory receptor SHPS-1|Macrophage fusion receptor|MyD-1 antigen|Signal-regulatory protein alpha-1|Signal-regulatory protein alpha-2|Signal-regulatory protein alpha-3|p84|Sirp-alpha-1|Sirp-alpha-2|Sirp-alpha-3
- BIT|MFR|MYD1|PTPNS1|SHPS1|SIRP|SIRPA|Tyrosine-protein phosphatase non-receptor type substrate 1|SHP substrate 1|SHPS-1|Brain Ig-like molecule with tyrosine-based activation motifs|CD172 antigen-like family member A|Inhibitory receptor SHPS-1|Macrophage fusion receptor|MyD-1 antigen|Signal-regulatory protein alpha-1|Signal-regulatory protein alpha-2|Signal-regulatory protein alpha-3|p84|Sirp-alpha-1|Sirp-alpha-2|Sirp-alpha-3
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SADF10.1
- SADF10.1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 54967 Da
- 54967 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- ICC/IF
- ICC/IF
Specifications
Frequently Bought Together