Anti-CD42a/GP-IX Mouse Monoclonal Antibody [clone: GR-P] (APC)
Catalog # ABCA_AB270643-10TE
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD42a
- Antigen Symbol:CD42a/GP-IX
- Clonality:Monoclonal
- Clone:GR-P
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:10 Tests
- Form:Liquid
- Antigen Synonyms:Platelet glycoprotein IX|GP-IX|GPIX|Glycoprotein 9|GP9
- Molecular Weight:19046 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Store at +4 °C. Store In the Dark.
- Shipping Temperature:+4 °C
- Immunogen:Cell preparation containing GP9 protein. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Size-exclusion chromatography
- Packaging:Vial
Specifications
About this item
APC Mouse monoclonal [GR-P] to CD42a/GP-IX.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD42a/GP-IX
Clonality: Monoclonal
Clone: GR-P
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human