Anti-CIKS/ACT1 Rabbit Polyclonal Antibody
Catalog # ABCA_AB137395-50UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:C6orf2
- Antigen Symbol:CIKS/ACT1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:C6orf4|C6orf5|C6orf6|TRAF3IP2|E3 ubiquitin ligase TRAF3IP2|Adapter protein CIKS|Connection to IKK and SAPK/JNK|E3 ubiquitin-protein ligase CIKS|Nuclear factor NF-kappa-B activator 1|TRAF3-interacting protein 2|ACT1
- Molecular Weight:64666 Da
- Storage Temperature:Shipped at +4 °C. Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
- Concentration:0.87 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human TRAF3IP2 aa 350 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to CIKS/ACT1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CIKS/ACT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human