Anti-IL-10RB/IL-10R2 Rabbit Monoclonal Antibody [clone: EPR21829-219]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR21829-219] to IL-10RB/IL-10R2 - BSA and Azide free (Detector).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: IL-10RB/IL-10R2
Clonality: Monoclonal
Clone: EPR21829-219
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB259506-1MG
- ABCA_AB259506-100U
- Antigen Symbol:
- IL-10RB/IL-10R2
- IL-10RB/IL-10R2
- Antigen Name:
- CDw210b
- CDw210b
- Antigen Synonyms:
- CRFB4|D21S58|D21S66|IL10RB|Interleukin-10 receptor subunit beta|IL-10 receptor subunit beta|IL-10R subunit beta|IL-10RB|Cytokine receptor class-II member 4|Cytokine receptor family 2 member 4|Interleukin-10 receptor subunit 2|CRF2-4|IL-10R subunit 2|IL-10R2
- CRFB4|D21S58|D21S66|IL10RB|Interleukin-10 receptor subunit beta|IL-10 receptor subunit beta|IL-10R subunit beta|IL-10RB|Cytokine receptor class-II member 4|Cytokine receptor family 2 member 4|Interleukin-10 receptor subunit 2|CRF2-4|IL-10R subunit 2|IL-10R2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR21829-219
- EPR21829-219
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 36995 Da
- 36995 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- sELISA
- sELISA
Specifications
Frequently Bought Together