Anti-GARS Mouse Monoclonal Antibody [clone: AT4E10]
Catalog # ABCA_AB282154-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:GARS1
- Antigen Symbol:GARS
- Clonality:Monoclonal
- Clone:AT4E10
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Glycine--tRNA ligase|Diadenosine tetraphosphate synthetase|Glycyl-tRNA synthetase|Glycyl-tRNA synthetase 1|Ap4A synthetase|GlyRS
- Molecular Weight:83166 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human GARS1 aa 1-300. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [AT4E10] to GARS.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GARS
Clonality: Monoclonal
Clone: AT4E10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human