Anti-ERGIC1 Rabbit Polyclonal Antibody
Catalog # ABCA_AB121582-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:ERGIC32
- Antigen Symbol:ERGIC1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:KIAA1181|HT034|Endoplasmic reticulum-Golgi intermediate compartment protein 1|ER-Golgi intermediate compartment 32 kDa protein|ERGIC-32
- Molecular Weight:32592 Da
- Storage Temperature:Upon delivery aliquot and store at –20 °C. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human ERGIC1 aa 100 - 200. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to ERGIC1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ERGIC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human