Anti-TEM1 Rabbit Polyclonal Antibody
ABCA_AB217535-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:CD248
- Antigen Symbol:TEM1
- Clonality:Polyclonal
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:CD164L1|Endosialin|Tumor endothelial marker 1
- Molecular Weight:80859 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Synthetic Peptide within Human CD248 aa 250-350 conjugated to Keyhole Limpet Haemocyanin. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein A Affinity purification
- Packaging:Vial
Rabbit polyclonal to TEM1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TEM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human