Anti-Kir2.1/KCNJ2 Rabbit Polyclonal Antibody
ABCA_AB65796-100UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:IRK1
- Antigen Symbol:Kir2.1/KCNJ2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:KCNJ2|Inward rectifier potassium channel 2|Cardiac inward rectifier potassium channel|Inward rectifier K(+) channel Kir2.1|IRK-1|hIRK1
- Storage Buffer:Whole serum
- Molecular Weight:48288 Da
- Storage Temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:ICC/IF, IHC-P
- Purification:Whole antiserum
- Packaging:Vial
Rabbit polyclonal to Kir2.1/KCNJ2.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Kir2.1/KCNJ2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat