Anti-Hsp70 Mouse Monoclonal Antibody [clone: 5A5]
Catalog # ABCA_AB2787-250UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:HSP72
- Antigen Symbol:Hsp70
- Clonality:Monoclonal
- Clone:5A5
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Mouse,Rat,Human,Dog,African green monkey
- Western Blot:Yes
- Size:250 μl
- Epitope:Epitope mapping with a panel of HSP 70 deletion mutants suggests that the epitope recognized is located between amino acids 122 - 264 of human HSP 70, a region that has been shown to be involved in ATP binding. This is the first monoclonal antibody reported to react with: 1) The ATP binding region of HSP 70. 2) An epitope in the amino terminus of HSP 70.
- Form:Liquid
- Antigen Synonyms:HSPA1|HSX70|HSPA1A|Heat shock 70 kDa protein 1A|Heat shock 70 kDa protein 1|Heat shock protein family A member 1A|HSP70-1|HSP70.1
- Storage Buffer:PBS
- Molecular Weight:70052 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Concentration:9.3 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human HSPA1A aa 100 - 300. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [5A5] to Hsp70.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Hsp70
Clonality: Monoclonal
Clone: 5A5
Conjugation:
Epitope: Epitope mapping with a panel of HSP 70 deletion mutants suggests that the epitope recognized is located between amino acids 122 - 264 of human HSP 70, a region that has been shown to be involved in ATP binding. This is the first monoclonal antibody reported to react with: 1) The ATP binding region of HSP 70. 2) An epitope in the amino terminus of HSP 70.
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat;Human;Dog;African green monkey