Anti-CD47 Rabbit Recombinant Multiclonal Antibody [clone: RM1014]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit recombinant multiclonal [RM1014] to CD47 - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD47
Clonality: Recombinant Multiclonal
Clone: RM1014
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB284138-100U
- ABCA_AB284138-1MG
- Antigen Symbol:
- CD47
- CD47
- Antigen Name:
- MER6
- MER6
- Antigen Synonyms:
- Leukocyte surface antigen CD47|Antigenic surface determinant protein OA3|Integrin-associated protein|Protein IAP
- Leukocyte surface antigen CD47|Antigenic surface determinant protein OA3|Integrin-associated protein|Protein IAP
- Antibody Type:
- Primary
- Primary
- Clone:
- RM1014
- RM1014
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 35214 Da
- 35214 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C.
- Store at +4 °C.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Specifications
Frequently Bought Together