Anti-PKA beta (catalytic subunit)+cAMP Protein Kinase Catalytic subunit (phospho T197) Rabbit Recombinant Multiclonal Antibody [clone: RP23040216]
ABCA_AB313487-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:PKACA
- Antigen Symbol:PKA beta (catalytic subunit)+cAMP Protein Kinase Catalytic subunit
- Clonality:Recombinant Multiclonal
- Clone:RP23040216
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:PRKACA|cAMP-dependent protein kinase catalytic subunit alpha|PKA C-alpha
- Modification:phospho T197
- Molecular Weight:40590 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human PRKACA phospho T197
- Tested Applications:ICC/IF
- Purification:Protein A affinity purification
- Packaging:Vial
Rabbit recombinant multiclonal [RP23040216] to PKA beta (catalytic subunit) (phospho T197) + cAMP Protein Kinase Catalytic subunit (phospho T197).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PKA beta (catalytic subunit)+cAMP Protein Kinase Catalytic subunit
Clonality: Recombinant Multiclonal
Clone: RP23040216
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human