Anti-CD200R Mouse Monoclonal Antibody [clone: OX108] (PE (Phycoerythrin))
Catalog # ABCA_AB33366-50TES
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CRTR2
- Antigen Symbol:CD200R
- Clonality:Monoclonal
- Clone:OX108
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Size:50 Tests
- Form:Liquid
- Antigen Synonyms:MOX2R|OX2R|UNQ2522/PRO6015|CD200R1|Cell surface glycoprotein CD200 receptor 1|CD200 cell surface glycoprotein receptor|Cell surface glycoprotein OX2 receptor 1
- Storage Buffer:PBS, BSA
- Molecular Weight:36620 Da
- Storage Temperature:Store at 4 °C
- Shipping Temperature:4 °C
- Immunogen:Recombinant Protein within Human CD200R1. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Protein G affinity purification
- Packaging:Vial
Specifications
About this item
PE Mouse monoclonal [OX108] to CD200R.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD200R
Clonality: Monoclonal
Clone: OX108
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human