Anti-HLA DQA2 Mouse Monoclonal Antibody [clone: F3.3]
Catalog # ABCA_AB288433-200U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:HLA-DXA
- Antigen Symbol:HLA DQA2
- Clonality:Monoclonal
- Clone:F3.3
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:200 µg
- Form:Liquid
- Antigen Synonyms:HLA-DQA2|DX alpha chain|HLA-DQA1|MHC class II DQA2
- Molecular Weight:28033 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [F3.3] to HLA DQA2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HLA DQA2
Clonality: Monoclonal
Clone: F3.3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human