Anti-PRKAR1B Mouse Polyclonal Antibody
Catalog # ABCA_AB167325-50UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:cAMP-dependent protein kinase type I-beta regulatory subunit
- Antigen Symbol:PRKAR1B
- Clonality:Polyclonal
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Molecular Weight:43073 Da
- Storage Temperature:Shipped at +4 °C. Upon delivery aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human cAMP-dependent protein kinase type I-beta regulatory subunit.
- Tested Applications:ICC/IF
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse polyclonal to PRKAR1B.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PRKAR1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human