Anti-Lamin A + Lamin C (phospho S22) Rabbit Monoclonal Antibody [clone: LaminACS22-CF12]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [LaminACS22-CF12] to Lamin A + Lamin C (phospho S22).
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Lamin A + Lamin C
Clonality: Monoclonal
Clone: LaminACS22-CF12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- ABCA_AB278643-20UL
- ABCA_AB278643-100U
- Antigen Symbol:
- Lamin A + Lamin C
- Lamin A + Lamin C
- Antigen Name:
- LMN1
- LMN1
- Antigen Synonyms:
- LMNA|Prelamin-A/C
- LMNA|Prelamin-A/C
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- LaminACS22-CF12
- LaminACS22-CF12
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Modification:
- phospho S22
- phospho S22
- Immunogen:
- Synthetic Peptide within Human LMNA phospho S22. The exact immunogen used to generate this antibody is proprietary information.
- Synthetic Peptide within Human LMNA phospho S22. The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 74139 Da
- 74139 Da
- Purification:
- Protein A/G affinity purification
- Protein A/G affinity purification
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together