Order Entry
United States
ContactUsLinkComponent
Anti-SLAM/CD150 Rabbit Monoclonal Antibody [clone: EPR22904-135]
Anti-SLAM/CD150 Rabbit Monoclonal Antibody [clone: EPR22904-135]
Supplier:  Abcam
New Product
Certificates
Anti-SLAM/CD150 Rabbit Monoclonal Antibody [clone: EPR22904-135]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB228978-100U
  • ABCA_AB228978-1ML
  • Antigen Symbol:
  • SLAM / CD150
  • SLAM / CD150
  • Antigen Name:
  • CD150
  • CD150
  • Antigen Synonyms:
  • Slam|Slamf1|Signaling lymphocytic activation molecule|SLAM family member 1
  • Slam|Slamf1|Signaling lymphocytic activation molecule|SLAM family member 1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR22904-135
  • EPR22904-135
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information.
  • The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 38094 Da
  • 38094 Da
  • Purification:
  • Protein A Affinity purification
  • Protein A Affinity purification
  • Storage Temperature:
  • Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
  • Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • Shipped at +4 °C
  • Shipped at +4 °C
  • Tested Applications:
  • IHC-P
  • IHC-P

Specifications

Frequently Bought Together