Order Entry
United States
ContactUsLinkComponent
Anti-Lysosomal acid lipase/LAL Rabbit Polyclonal Antibody
Anti-Lysosomal acid lipase/LAL Rabbit Polyclonal Antibody
Supplier:  Abcam
New Product
Certificates
Anti-Lysosomal acid lipase/LAL Rabbit Polyclonal Antibody
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB154356-100U
  • ABCA_AB154356-25UL
  • Antigen Symbol:
  • Lysosomal acid lipase/LAL
  • Lysosomal acid lipase/LAL
  • Antigen Name:
  • Lysosomal acid lipase/cholesteryl ester hydrolase
  • Lysosomal acid lipase/cholesteryl ester hydrolase
  • Antigen Synonyms:
  • Acid cholesteryl ester hydrolase|LAL|Cholesteryl esterase|Diacylglycerol lipase|Lipase A|Sterol esterase|Triacylglycerol ester hydrolase|Triacylglycerol lipase|LIPA
  • Acid cholesteryl ester hydrolase|LAL|Cholesteryl esterase|Diacylglycerol lipase|Lipase A|Sterol esterase|Triacylglycerol ester hydrolase|Triacylglycerol lipase|LIPA
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Clone:
  • 0
  • 0
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Fragment Protein within Human LIPA aa 150 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
  • Recombinant Fragment Protein within Human LIPA aa 150 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
  • Form:
  • Liquid
  • Liquid
  • Molecular Weight:
  • 45419 Da
  • 45419 Da
  • Purification:
  • Immunogen Affinity purification
  • Immunogen Affinity purification
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage Temperature:
  • Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
  • Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • Shipped at +4 °C
  • Shipped at +4 °C

Specifications

Frequently Bought Together