Anti-MDM2 Mouse Monoclonal Antibody [clone: SMP 14]
Catalog # ABCA_AB3110-100UG
Supplier: Abcam
New Product
Specifications
- Size:100 µg
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [SMP 14] to MDM2.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: SMP 14
Conjugation:
Epitope: Amino acids 154 - 167.
Host: Mouse
Isotype: IgG1
Reactivity: Human