Anti-Melanoma Associated Antigen 100+/7 kDa Mouse Monoclonal Antibody [clone: NKI/beteb]
Catalog # ABCA_AB34165-1ML
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:MAA
- Antigen Symbol:Melanoma Associated Antigen 100+ / 7 kDa
- Clonality:Monoclonal
- Clone:NKI/beteb
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Size:1 ml
- Form:Liquid
- Storage Buffer:PBS, BSA
- Storage Temperature:Store at 4 °C (stable for up to 12 months). Upon delivery aliquot. Store at +4 °C.
- Concentration:0.1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Tissue preparation containing Melanoma Associated Antigen 100+/7 kDa protein. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein G Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [NKI/beteb] to Melanoma Associated Antigen 100+ / 7 kDa.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Melanoma Associated Antigen 100+ / 7 kDa
Clonality: Monoclonal
Clone: NKI/beteb
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human