Order Entry
United States
ContactUsLinkComponent
Anti-Mus81 Mouse Monoclonal Antibody [clone: MTA30 2G10/3]
Anti-Mus81 Mouse Monoclonal Antibody [clone: MTA30 2G10/3]
  ABCA_AB14387-100UG
 :  Abcam
New Product
Anti-Mus81 Mouse Monoclonal Antibody [clone: MTA30 2G10/3]
  ABCA_AB14387-100UG
 :  Abcam
 :  AB14387-100UG
New Product

 

  • Size:
    100 µg
  • Packaging:
    Vial

 

 

Mouse monoclonal [MTA30 2G10/3] to Mus81.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Mus81
Clonality: Monoclonal
Clone: MTA30 2G10/3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human