Anti-HLA-DQB1+HLA Class II DRB1+HLA-DPB1 Mouse Monoclonal Antibody [clone: Bu26]
Catalog # ABCA_AB316364-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:HLA-DP1B
- Antigen Symbol:HLA-DQB1+HLA Class II DRB1+HLA-DPB1
- Clonality:Monoclonal
- Clone:Bu26
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:HLA-DPB1|MHC class II antigen DPB1
- Molecular Weight:29159 Da
- Storage Temperature:Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Store at –20 °C. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Native Protein within Human HLA-DPB1.
- Tested Applications:ICC/IF
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [Bu26] to HLA-DQB1 + HLA Class II DRB1 + HLA-DPB1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HLA-DQB1+HLA Class II DRB1+HLA-DPB1
Clonality: Monoclonal
Clone: Bu26
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human