Anti-Tcl1 Rabbit Monoclonal Antibody [clone: EPR3949]
: Abcam
New Product
Rabbit monoclonal [EPR3949] to Tcl1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Tcl1
Clonality: Monoclonal
Clone: EPR3949
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB108978-1ML
- ABCA_AB108978-100U
- Antigen Symbol:
- Tcl1
- Tcl1
- Antigen Name:
- TCL1A
- TCL1A
- Antigen Synonyms:
- T-cell leukemia/lymphoma protein 1A|Oncogene TCL-1|Protein p14 TCL1|Oncogene TCL1
- T-cell leukemia/lymphoma protein 1A|Oncogene TCL-1|Protein p14 TCL1|Oncogene TCL1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR3949
- EPR3949
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 13460 Da
- 13460 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Buffer:
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- Storage Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Frequently Bought Together