Anti-IP3 receptor Rabbit Monoclonal Antibody [clone: EPR4537]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR4537] to IP3 receptor.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: IP3 receptor
Clonality: Monoclonal
Clone: EPR4537
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB108517-100U
- ABCA_AB108517-1ML
- Antigen Symbol:
- IP3 receptor
- IP3 receptor
- Antigen Name:
- INSP3R1
- INSP3R1
- Antigen Synonyms:
- ITPR1|IP3 receptor isoform 1|IP3R 1|Type 1 InsP3 receptor
- ITPR1|IP3 receptor isoform 1|IP3R 1|Type 1 InsP3 receptor
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR4537
- EPR4537
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 313929 Da
- 313929 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Buffer:
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- Storage Temperature:
- Store at −20 °C. Stable for 12 months at −20 °C.
- Store at −20 °C. Stable for 12 months at −20 °C.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P
Specifications
Frequently Bought Together