Order Entry
United States
ContactUsLinkComponent
Anti-COX6A2 Mouse Monoclonal Antibody [clone: 4H2A5]
Anti-COX6A2 Mouse Monoclonal Antibody [clone: 4H2A5]
Catalog # ABCA_AB110264-250U
Supplier:  Abcam
New Product
Anti-COX6A2 Mouse Monoclonal Antibody [clone: 4H2A5]
Catalog # ABCA_AB110264-250U
Supplier:  Abcam
Supplier Number:  AB110264-250UG
New Product

Specifications

  • Size:
    250 µg
  • Packaging:
    Vial

Specifications

About this item

Mouse monoclonal [4H2A5] to COX6A2.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: COX6A2
Clonality: Monoclonal
Clone: 4H2A5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine