Anti-eIF4A2 Mouse Polyclonal Antibody
Catalog # ABCA_AB194471-50UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:DDX2B
- Antigen Symbol:eIF4A2
- Clonality:Polyclonal
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:EIF4F|Eukaryotic initiation factor 4A-II|eIF-4A-II|eIF4A-II|ATP-dependent RNA helicase eIF4A-2
- Molecular Weight:46402 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human EIF4A2 aa 1-100. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Whole antiserum
- Packaging:Vial
Specifications
About this item
Mouse polyclonal to eIF4A2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: eIF4A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human