Anti-Argonaute-2 Rabbit Monoclonal Antibody [clone: EPR10410]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR10410] to Argonaute-2 - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Argonaute-2
Clonality: Monoclonal
Clone: EPR10410
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB249284-100U
- ABCA_AB249284-1MG
- Antigen Symbol:
- Argonaute-2
- Argonaute-2
- Antigen Name:
- EIF2C2
- EIF2C2
- Antigen Synonyms:
- AGO2|Protein argonaute-2|Argonaute2|hAgo2|Argonaute RISC catalytic component 2|Eukaryotic translation initiation factor 2C 2|PAZ Piwi domain protein|Protein slicer|eIF-2C 2|eIF2C 2|PPD
- AGO2|Protein argonaute-2|Argonaute2|hAgo2|Argonaute RISC catalytic component 2|Eukaryotic translation initiation factor 2C 2|PAZ Piwi domain protein|Protein slicer|eIF-2C 2|eIF2C 2|PPD
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR10410
- EPR10410
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 97208 Da
- 97208 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C. Do not freeze.
- Store at +4 °C. Do not freeze.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together