Anti-CD32B+CD32A Mouse Monoclonal Antibody [clone: AT10] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCA_AB30356-100UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CD32
- Antigen Symbol:CD32B+CD32A
- Clonality:Monoclonal
- Clone:AT10
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:FCG2|IGFR2|FCGR2B|Low affinity immunoglobulin gamma Fc region receptor II-b|IgG Fc receptor II-b|CDw32|Fc-gamma RII-b|Fc-gamma-RIIb|FcRII-b
- Storage Buffer:PBS, BSA
- Molecular Weight:34044 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:Flow Cyt (Intra), ICC/IF
- Purification:Protein G affinity purification
- Packaging:Vial
Specifications
About this item
FITC Mouse monoclonal [AT10] to CD32B + CD32A.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD32B+CD32A
Clonality: Monoclonal
Clone: AT10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human