Anti-RUNX1 / AML1 Rabbit Polyclonal Antibody
ABCA_AB23980-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:AML1
- Antigen Symbol:RUNX1 / AML1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:CBFA2|RUNX1|Runt-related transcription factor 1|Acute myeloid leukemia 1 protein|Core-binding factor subunit alpha-2|Oncogene AML-1|Polyomavirus enhancer-binding protein 2 alpha B subunit|SL3-3 enhancer factor 1 alpha B subunit|SL3/AKV core-binding factor alpha B subunit|CBF-alpha-2|PEA2-alpha B|PEBP2-alpha B
- Storage Buffer:PBS, BSA
- Molecular Weight:48737 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Concentration:0.7 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to RUNX1 / AML1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: RUNX1 / AML1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human