Anti-GABA A Receptor beta 3/GABRB3 Mouse Monoclonal Antibody [clone: N87/25]
ABCA_AB98968-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Gamma-aminobutyric acid receptor subunit beta-3
- Antigen Symbol:GABA A Receptor beta 3/GABRB3
- Clonality:Monoclonal
- Clone:N87/25
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:GABA(A) receptor subunit beta-3|GABAAR subunit beta-3|GABRB3
- Storage Buffer:PBS, glycerol
- Molecular Weight:54116 Da
- Storage Temperature:Store at −20 °C. Stable for 12 months at −20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Protein within Mouse Gabrb3 aa 350-450. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-Fr, IHC-P
- Purification:Protein G affinity purification
- Packaging:Vial
Mouse monoclonal [N87/25] to GABA A Receptor beta 3/GABRB3.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GABA A Receptor beta 3/GABRB3
Clonality: Monoclonal
Clone: N87/25
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat