Anti-Pyrin Rabbit Monoclonal Antibody [clone: EPR18675]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR18675] to Pyrin - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Pyrin
Clonality: Monoclonal
Clone: EPR18675
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- ABCA_AB223132-1MG
- ABCA_AB223132-100U
- Antigen Symbol:
- Pyrin
- Pyrin
- Antigen Name:
- Mefv
- Mefv
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR18675
- EPR18675
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 86409 Da
- 86409 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IP
- IP
Specifications
Frequently Bought Together