Anti-GOSR1/GS28 Rabbit Monoclonal Antibody [clone: EP1768Y]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EP1768Y] to GOSR1/GS28.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GOSR1/GS28
Clonality: Monoclonal
Clone: EP1768Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- ABCA_AB53288-100UL
- ABCA_AB53288-1ML
- Antigen Symbol:
- GOSR1/GS28
- GOSR1/GS28
- Antigen Name:
- GS28
- GS28
- Antigen Synonyms:
- GOSR1|Golgi SNAP receptor complex member 1|28 kDa Golgi SNARE protein|28 kDa cis-Golgi SNARE p28|GOS-28
- GOSR1|Golgi SNAP receptor complex member 1|28 kDa Golgi SNARE protein|28 kDa cis-Golgi SNARE p28|GOS-28
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EP1768Y
- EP1768Y
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 28613 Da
- 28613 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Buffer:
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- BSA, glycerol, tissue culture supernatant, sodium chloride, tris glycine
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Stable for 12 months at −20 °C.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Stable for 12 months at −20 °C.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
Specifications
Frequently Bought Together