Anti-KPRP Rabbit Polyclonal Antibody
ABCA_AB122201-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:C1orf45
- Antigen Symbol:KPRP
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Keratinocyte proline-rich protein|hKPRP
- Molecular Weight:64136 Da
- Storage Temperature:Store at −20 or −80 °C. Avoid freeze/thaw cycle. Store undiluted.
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human KPRP. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen affinity purification
- Packaging:Vial
Rabbit polyclonal to KPRP.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: KPRP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human