Anti-Cytokeratin 6 Rabbit Monoclonal Antibody [clone: EPR1602Y]
: Abcam
New Product
Rabbit monoclonal [EPR1602Y] to Cytokeratin 6.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cytokeratin 6
Clonality: Monoclonal
Clone: EPR1602Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB93279-1ML
- ABCA_AB93279-100UL
- Antigen Symbol:
- Cytokeratin 6
- Cytokeratin 6
- Antigen Name:
- KRT6E
- KRT6E
- Antigen Synonyms:
- KRT6C|Cytokeratin-6C|Cytokeratin-6E|Keratin K6h|Keratin-6C|Type-II keratin Kb12|CK-6C|CK-6E|K6C
- KRT6C|Cytokeratin-6C|Cytokeratin-6E|Keratin K6h|Keratin-6C|Type-II keratin Kb12|CK-6C|CK-6E|K6C
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR1602Y
- EPR1602Y
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 60025 Da
- 60025 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Buffer:
- BSA, Glycerol, Sodium chloride, Tris glycine
- BSA, Glycerol, Sodium chloride, Tris glycine
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Stable for 12 months at –20 °C.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Stable for 12 months at –20 °C.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Frequently Bought Together