Anti-ribosomal RNA Mouse Monoclonal Antibody [clone: Y10B]
ABCA_AB171119-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Symbol:ribosomal RNA
- Clonality:Monoclonal
- Clone:Y10B
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG3
- Size:100 µg
- Form:Liquid
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Tested Applications:ICC/IF
- Purification:Protein G Affinity purification
- Packaging:Vial
Mouse monoclonal [Y10B] to ribosomal RNA.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ribosomal RNA
Clonality: Monoclonal
Clone: Y10B
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: