Anti-LRSAM1 Mouse Polyclonal Antibody
Catalog # ABCA_AB73113-50UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:TAL
- Antigen Symbol:LRSAM1
- Clonality:Polyclonal
- Host:Mouse
- Isotype:IgG
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Antigen Synonyms:UNQ6496/PRO21356|E3 ubiquitin-protein ligase LRSAM1|Leucine-rich repeat and sterile alpha motif-containing protein 1|RING-type E3 ubiquitin transferase LRSAM1|Tsg101-associated ligase|hTAL
- Storage Buffer:PBS
- Molecular Weight:83594 Da
- Storage Temperature:Upon delivery aliquot and Store at –20 or –80 °C. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human LRSAM1.
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse polyclonal to LRSAM1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: LRSAM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human