Anti-Angiopoietin 2/ANG2 Rabbit Monoclonal Antibody [clone: 1F10]
Catalog # ABCA_AB302883-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Angiopoietin-2
- Antigen Symbol:Angiopoietin 2/ANG2
- Clonality:Monoclonal
- Clone:1F10
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:ANG-2|ANGPT2
- Molecular Weight:56919 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein A Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit monoclonal [1F10] to Angiopoietin 2/ANG2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Angiopoietin 2/ANG2
Clonality: Monoclonal
Clone: 1F10
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human