Anti-SRPR alpha Rabbit Polyclonal Antibody
Supplier: Abcam
New Product
Certificates
About this item
Rabbit polyclonal to SRPR alpha.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SRPR alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Specifications
- Catalog No:
- ABCA_AB228625-100U
- ABCA_AB228625-25UL
- Antigen Symbol:
- SRPR alpha
- SRPR alpha
- Antigen Name:
- SRPR
- SRPR
- Antigen Synonyms:
- SRPRA|Signal recognition particle receptor subunit alpha|SR-alpha|Docking protein alpha|DP-alpha
- SRPRA|Signal recognition particle receptor subunit alpha|SR-alpha|Docking protein alpha|DP-alpha
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Mouse, Human
- Mouse, Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Fragment Protein within Human SRPRA. The exact immunogen used to generate this antibody is proprietary information.
- Recombinant Fragment Protein within Human SRPRA. The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 69811 Da
- 69811 Da
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P
Specifications
Frequently Bought Together