Anti-Transglutaminase 2 Rabbit Monoclonal Antibody [clone: EP2957]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EP2957] to Transglutaminase 2.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Transglutaminase 2
Clonality: Monoclonal
Clone: EP2957
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB109200-1ML
- ABCA_AB109200-100U
- Antigen Symbol:
- Transglutaminase 2
- Transglutaminase 2
- Antigen Name:
- Protein-glutamine gamma-glutamyltransferase 2
- Protein-glutamine gamma-glutamyltransferase 2
- Antigen Synonyms:
- Erythrocyte transglutaminase|Heart G alpha(h)|Isopeptidase TGM2|Protein G alpha(h)|Protein-glutamine deamidase TGM2|Protein-glutamine dopaminyltransferase TGM2|Protein-glutamine histaminyltransferase TGM2|Protein-glutamine noradrenalinyltransferase TGM2|Protein-glutamine serotonyltransferase TGM2|Tissue transglutaminase|Transglutaminase C|Transglutaminase H|Transglutaminase II|Transglutaminase-2|hhG alpha(h)|G(h)|tTG|tTgase|TG(C)|TGC|TGase C|TGase H|TGase II|TG2|TGase-2|hTG2|TGM2
- Erythrocyte transglutaminase|Heart G alpha(h)|Isopeptidase TGM2|Protein G alpha(h)|Protein-glutamine deamidase TGM2|Protein-glutamine dopaminyltransferase TGM2|Protein-glutamine histaminyltransferase TGM2|Protein-glutamine noradrenalinyltransferase TGM2|Protein-glutamine serotonyltransferase TGM2|Tissue transglutaminase|Transglutaminase C|Transglutaminase H|Transglutaminase II|Transglutaminase-2|hhG alpha(h)|G(h)|tTG|tTgase|TG(C)|TGC|TGase C|TGase H|TGase II|TG2|TGase-2|hTG2|TGM2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EP2957
- EP2957
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 77329 Da
- 77329 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Buffer:
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- Storage Temperature:
- Shipped at +4 °C. Store at –20 °C. Stable for 12 months at –20 °C.
- Shipped at +4 °C. Store at –20 °C. Stable for 12 months at –20 °C.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P
Specifications
Frequently Bought Together