Anti-Glutathione Peroxidase 4 Rabbit Polyclonal Antibody
ABCA_AB41787-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Phospholipid hydroperoxide glutathione peroxidase GPX4
- Antigen Symbol:Glutathione Peroxidase 4
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:PHGPx|GPx-4|GSHPx-4|GPX4
- Storage Buffer:PBS, BSA
- Molecular Weight:22175 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to Glutathione Peroxidase 4.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Glutathione Peroxidase 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human