Anti-Sarcomeric Alpha Actinin Rabbit Polyclonal Antibody
Catalog # ABCA_AB72592-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Alpha-actinin-2
- Antigen Symbol:Sarcomeric Alpha Actinin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Alpha-actinin skeletal muscle isoform 2|F-actin cross-linking protein|ACTN2
- Storage Buffer:PBS, Glycerol, Sodium chloride
- Molecular Weight:103854 Da
- Storage Temperature:Store at –20 °C. Stable for 12 months at –20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human ACTN2. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF, IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Sarcomeric Alpha Actinin.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Sarcomeric Alpha Actinin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human