Anti-YB1 Mouse Monoclonal Antibody [clone: YBX1/2430]
Catalog # ABCA_AB268094-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:NSEP1
- Antigen Symbol:YB1
- Clonality:Monoclonal
- Clone:YBX1/2430
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:YBX1|Y-box-binding protein 1|YB-1|CCAAT-binding transcription factor I subunit A|DNA-binding protein B|Enhancer factor I subunit A|Nuclease-sensitive element-binding protein 1|Y-box transcription factor|CBF-A|DBPB|EFI-A
- Molecular Weight:35924 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:0.2 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Human YBX1
- Tested Applications:IHC-P, Protein Array
- Purification:Protein A/G affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [YBX1/2430] to YB1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: YB1
Clonality: Monoclonal
Clone: YBX1/2430
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human