Anti-Mannose Receptor Rabbit Monoclonal Antibody [clone: EPR22489-7] (Alexa Fluor® 488)
: Abcam
New Product
Alexa Fluor® 488 Rabbit monoclonal [EPR22489-7] to Mannose Receptor.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Mannose Receptor
Clonality: Monoclonal
Clone: EPR22489-7
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB313398-10UL
- ABCA_AB313398-100U
- Antigen Symbol:
- Mannose Receptor
- Mannose Receptor
- Antigen Name:
- CD206
- CD206
- Antigen Synonyms:
- CLEC13D|CLEC13DL|MRC1L1|MRC1|Macrophage mannose receptor 1|MMR|C-type lectin domain family 13 member D|C-type lectin domain family 13 member D-like|Human mannose receptor|Macrophage mannose receptor 1-like protein 1|hMR
- CLEC13D|CLEC13DL|MRC1L1|MRC1|Macrophage mannose receptor 1|MMR|C-type lectin domain family 13 member D|C-type lectin domain family 13 member D-like|Human mannose receptor|Macrophage mannose receptor 1-like protein 1|hMR
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 488
- Alexa Fluor® 488
- Clone:
- EPR22489-7
- EPR22489-7
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 166012 Da
- 166012 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage Temperature:
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle. Store in the dark.
- Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle. Store in the dark.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P
Frequently Bought Together